Assay Details
| Gene Symbol: CMTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711555.3 | XP_002711601.1 | 3 - 4 | 531 | 72 | |
| XM_017342271.1 | XP_017197760.1 | 4 - 5 | 605 | 72 | ||
More Information
Set Membership: |
|

CMTM3
Genomic Map