Assay Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273208.2 | XP_008271430.1 | 4 - 5 | 654 | 60 | |
| XM_008273210.2 | XP_008271432.1 | 3 - 4 | 649 | 60 | ||
| XM_017350020.1 | XP_017205509.1 | 3 - 4 | 655 | 60 | ||
| XM_017350021.1 | XP_017205510.1 | 4 - 5 | 677 | 60 | ||
| XM_017350022.1 | XP_017205511.1 | 2 - 3 | 589 | 60 | ||
More Information
Set Membership: |
|

ARHGAP36
Genomic Map