Assay Details
| Gene Symbol: RABGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722278.3 | XP_002722324.1 | 1194 | 67 | ||
| XM_008249650.2 | XP_008247872.1 | 1197 | 67 | |||
| XM_017338225.1 | XP_017193714.1 | 648 | 67 | |||
More Information
Set Membership: |
|

RABGEF1
Genomic Map