Assay Details
| Gene Symbol: LOC100344750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267611.2 | XP_008265833.1 | 9 - 10 | 1285 | 101 | |
| XM_008267612.2 | XP_008265834.1 | 9 - 10 | 1182 | 101 | ||
| XM_017347410.1 | XP_017202899.1 | 9 - 10 | 1287 | 101 | ||
More Information
Set Membership: |
|

LOC100344750
Genomic Map