Assay Details
| Gene Symbol: TAPBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259728.2 | XP_008257950.1 | 4 - 5 | 948 | 75 | |
| XM_008259729.2 | XP_008257951.1 | 4 - 5 | 1278 | 75 | ||
| XM_017343403.1 | XP_017198892.1 | 4 - 5 | 1283 | 75 | ||
More Information
Set Membership: |
|

TAPBPL
Genomic Map