Assay Details
| Gene Symbol: TENT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261867.2 | XP_008260089.1 | 11 - 12 | 1635 | 61 | |
| XM_008261869.2 | XP_008260091.1 | 11 - 12 | 1623 | 61 | ||
More Information
Set Membership: |
|

TENT2
Genomic Map