Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256762.2 | XP_008254984.1 | 4 - 5 | 634 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM38B| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256762.2 | XP_008254984.1 | 4 - 5 | 634 | 65 | |
Set Membership: |
|