Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271629.2 | XP_008269851.1 | 8 - 9 | 844 | 56 | |
| XM_008271630.2 | XP_008269852.1 | 8 - 9 | 737 | 56 | ||
| XM_008271632.2 | XP_008269854.1 | 7 - 8 | 640 | 56 | ||
More Information
Set Membership: |
|

DBF4B
Genomic Map