Assay Details
| Gene Symbol: FUT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345287.1 | XP_017200776.1 | 388 | 178 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017345287.1 transcripts.
Set Membership: |
|

FUT9
Genomic Map