Assay Details
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008269856.2 | XP_008268078.1 | 6 - 7 | 887 | 66 | |
XM_008269857.2 | XP_008268079.1 | 6 - 7 | 1045 | 66 | ||
XM_008269858.2 | XP_008268080.1 | 6 - 7 | 885 | 66 | ||
XM_008269859.2 | XP_008268081.1 | 6 - 7 | 902 | 66 | ||
XM_008269860.2 | XP_008268082.1 | 6 - 7 | 906 | 66 | ||
XM_008269861.2 | XP_008268083.1 | 6 - 7 | 902 | 66 | ||
XM_008269862.2 | XP_008268084.1 | 4 - 5 | 480 | 66 | ||
XM_017348678.1 | XP_017204167.1 | 4 - 5 | 744 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
