Assay Details
| Gene Symbol: LCA5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723243.3 | XP_002723289.1 | 1078 | 87 | ||
| XM_008251478.2 | XP_008249700.1 | 1225 | 87 | |||
| XM_008251479.2 | XP_008249701.1 | 3296 | 87 | |||
| XM_008251480.2 | XP_008249702.1 | 1225 | 87 | |||
More Information
Set Membership: |
|

LCA5L
Genomic Map