Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722224.3 | XP_002722270.1 | 855 | 61 | ||
| XM_008249535.2 | XP_008247757.1 | 739 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIN37| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722224.3 | XP_002722270.1 | 855 | 61 | ||
| XM_008249535.2 | XP_008247757.1 | 739 | 61 | |||
Set Membership: |
|