Assay Details
| Gene Symbol: C4H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256400.1 | XP_008254622.1 | 1 - 2 | 90 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008256400.1 transcripts.
Set Membership: |
|

C4H12orf54
Genomic Map