Assay Details
| Gene Symbol: C4H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256400.1 | XP_008254622.1 | 6 - 7 | 341 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H12orf54| Gene Symbol: C4H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256400.1 | XP_008254622.1 | 6 - 7 | 341 | 66 | |
Set Membership: |
|