Assay Details
| Gene Symbol: SYCP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344793.1 | XP_017200282.1 | 1 - 2 | 133 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYCP2L| Gene Symbol: SYCP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344793.1 | XP_017200282.1 | 1 - 2 | 133 | 80 | |
Set Membership: |
|