Assay Details
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341363.1 | XP_017196852.1 | 13 - 14 | 1964 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCMDC2| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341363.1 | XP_017196852.1 | 13 - 14 | 1964 | 69 | |
Set Membership: |
|