Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350352.1 | XP_017205841.1 | 212 | 59 | ||
| XM_017350353.1 | XP_017205842.1 | 213 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STAR| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350352.1 | XP_017205841.1 | 212 | 59 | ||
| XM_017350353.1 | XP_017205842.1 | 213 | 59 | |||
Set Membership: |
|