Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261621.2 | XP_008259843.1 | 3 - 4 | 801 | 83 | |
| XM_008261622.2 | XP_008259844.1 | 2 - 3 | 292 | 83 | ||
More Information
Set Membership: |
|

KBTBD2
Genomic Map