Assay Details
| Gene Symbol: ADNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252907.2 | XP_008251129.1 | 2776 | 93 | ||
| XM_017340084.1 | XP_017195573.1 | 1108 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008252907.2, XM_017340084.1 transcripts.
Set Membership: |
|

ADNP2
Genomic Map