Assay Details
| Gene Symbol: COQ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263308.2 | XP_008261530.1 | 6 - 7 | 908 | 80 | |
| XM_017345298.1 | XP_017200787.1 | 6 - 7 | 867 | 80 | ||
More Information
Set Membership: |
|

COQ3
Genomic Map