Assay Details
| Gene Symbol: LOC100359146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264431.2 | XP_008262653.2 | 2 - 3 | 239 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100359146| Gene Symbol: LOC100359146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264431.2 | XP_008262653.2 | 2 - 3 | 239 | 70 | |
Set Membership: |
|