Assay Details
Gene Symbol: PCED1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710912.3 | XP_002710958.2 | 4 - 5 | 675 | 91 | |
XM_008256237.2 | XP_008254459.1 | 4 - 5 | 544 | 91 | ||
XM_008256238.2 | XP_008254460.1 | 4 - 5 | 725 | 91 | ||
XM_008256240.2 | XP_008254462.1 | 4 - 5 | 542 | 91 | ||
XM_017341730.1 | XP_017197219.1 | 4 - 5 | 842 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
