Assay Details
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251108.1 | XP_008249330.1 | 524 | 55 | ||
| XM_017339016.1 | XP_017194505.1 | 524 | 55 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POC5| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251108.1 | XP_008249330.1 | 524 | 55 | ||
| XM_017339016.1 | XP_017194505.1 | 524 | 55 | |||
Set Membership: |
|