Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265477.1 | XP_008263699.1 | 1 - 2 | 2659 | 95 | |
| XM_008265480.2 | XP_008263702.1 | 1 - 2 | 2659 | 95 | ||
| XM_008265481.2 | XP_008263703.1 | 1 - 2 | 2659 | 95 | ||
| XM_017346411.1 | XP_017201900.1 | 1 - 2 | 2659 | 95 | ||
| XM_017346412.1 | XP_017201901.1 | 1 - 2 | 2659 | 95 | ||
| XM_017346413.1 | XP_017201902.1 | 1 - 2 | 2659 | 95 | ||
More Information
Set Membership: |
|

SLFNL1
Genomic Map