Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264954.2 | XP_008263176.1 | 8 - 9 | 1031 | 97 | |
| XM_008264956.2 | XP_008263178.1 | 8 - 9 | 1031 | 97 | ||
| XM_008264958.2 | XP_008263180.1 | 8 - 9 | 1031 | 97 | ||
More Information
Set Membership: |
|

DNAI3
Genomic Map