Assay Details
| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261735.2 | XP_008259957.1 | 2 - 3 | 1528 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008261735.2 transcripts.
Set Membership: |
|

TMBIM7
Genomic Map