Assay Details
| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261735.2 | XP_008259957.1 | 5 - 6 | 1926 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMBIM7| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261735.2 | XP_008259957.1 | 5 - 6 | 1926 | 78 | |
Set Membership: |
|