Assay Details
| Gene Symbol: LOC100345389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339304.1 | XP_017194793.1 | 162 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017339304.1 transcripts.
Set Membership: |
|

LOC100345389
Genomic Map