Assay Details
| Gene Symbol: C4H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256734.2 | XP_008254956.1 | 7 - 8 | 1531 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H12orf56| Gene Symbol: C4H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256734.2 | XP_008254956.1 | 7 - 8 | 1531 | 69 | |
Set Membership: |
|