Assay Details
| Gene Symbol: TMEM217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262852.2 | XP_008261074.1 | 3 - 4 | 188 | 82 | |
| XM_008262853.2 | XP_008261075.1 | 3 - 4 | 201 | 82 | ||
| XM_017345083.1 | XP_017200572.1 | 3 - 4 | 440 | 82 | ||
| XM_017345084.1 | XP_017200573.1 | 3 - 4 | 398 | 82 | ||
More Information
Set Membership: |
|

TMEM217
Genomic Map