Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719421.3 | XP_002719467.1 | 6 - 7 | 963 | 68 | |
| XM_017349218.1 | XP_017204707.1 | 5 - 6 | 832 | 68 | ||
More Information
Set Membership: |
|

TMEM106A
Genomic Map