Assay Details
| Gene Symbol: TIGD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267319.2 | XP_008265541.1 | 2 - 3 | 338 | 91 | |
| XM_008267322.2 | XP_008265544.1 | 2 - 3 | 269 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008267319.2, XM_008267322.2 transcripts.
Set Membership: |
|

TIGD4
Genomic Map