Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716403.3 | XP_002716449.1 | 4 - 5 | 931 | 71 | |
| XM_008266716.2 | XP_008264938.1 | 4 - 5 | 703 | 71 | ||
| XM_017347020.1 | XP_017202509.1 | 4 - 5 | 681 | 71 | ||
More Information
Set Membership: |
|

RNF168
Genomic Map