Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270858.2 | XP_008269080.1 | 11 - 12 | 2370 | 60 | |
| XM_017348990.1 | XP_017204479.1 | 10 - 11 | 2181 | 60 | ||
| XM_017348991.1 | XP_017204480.1 | 10 - 11 | 2125 | 60 | ||
| XM_017348992.1 | XP_017204481.1 | 9 - 10 | 1908 | 60 | ||
| XM_017348993.1 | XP_017204482.1 | 10 - 11 | 2127 | 60 | ||
More Information
Set Membership: |
|

KIAA0753
Genomic Map