Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720330.3 | XP_002720376.1 | 10 - 11 | 2370 | 78 | |
| XM_017349834.1 | XP_017205323.1 | 9 - 10 | 3281 | 78 | ||
More Information
Set Membership: |
|

RBMX
Genomic Map