Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341184.1 | XP_017196673.1 | 3 - 4 | 1138 | 74 | |
| XM_017341185.1 | XP_017196674.1 | 3 - 4 | 1595 | 74 | ||
| XM_017341186.1 | XP_017196675.1 | 2 - 3 | 799 | 74 | ||
More Information
Set Membership: |
|

SLC26A2
Genomic Map