Assay Details
| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343482.1 | XP_017198971.1 | 2 - 3 | 308 | 84 | |
| XM_017343483.1 | XP_017198972.1 | 2 - 3 | 305 | 84 | ||
More Information
Set Membership: |
|

LRRC63
Genomic Map