Assay Details
| Gene Symbol: ARL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273134.2 | XP_008271356.1 | 2 - 3 | 516 | 67 | |
| XM_017349890.1 | XP_017205379.1 | 2 - 3 | 517 | 67 | ||
| XM_017349891.1 | XP_017205380.1 | 2 - 3 | 1087 | 67 | ||
| XM_017349892.1 | XP_017205381.1 | 2 - 3 | 1089 | 67 | ||
More Information
Set Membership: |
|

ARL13A
Genomic Map