Assay Details
| Gene Symbol: GJA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715651.3 | XP_002715697.1 | 1 | 202 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002715651.3 transcripts.
Set Membership: |
|

GJA8
Genomic Map