Assay Details
| Gene Symbol: PGAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258964.2 | XP_008257186.1 | 1 - 2 | 89 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008258964.2 transcripts.
Set Membership: |
|

PGAP4
Genomic Map