Assay Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345053.1 | XP_017200542.1 | 18 - 19 | 2762 | 64 | |
| XM_017345054.1 | XP_017200543.1 | 18 - 19 | 2762 | 64 | ||
| XM_017345055.1 | XP_017200544.1 | 17 - 18 | 2699 | 64 | ||
| XM_017345056.1 | XP_017200545.1 | 18 - 19 | 2762 | 64 | ||
| XM_017345057.1 | XP_017200546.1 | 18 - 19 | 2762 | 64 | ||
| XM_017345058.1 | XP_017200547.1 | 19 - 20 | 2489 | 64 | ||
More Information
Set Membership: |
|

ANKS1A
Genomic Map