Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718935.3 | XP_002718981.2 | 1 - 2 | 187 | 68 | |
| XM_008271045.2 | XP_008269267.1 | 1 - 2 | 226 | 68 | ||
| XM_008271046.2 | XP_008269268.1 | 3 - 4 | 424 | 68 | ||
| XM_017349076.1 | XP_017204565.1 | 1 - 2 | 187 | 68 | ||
More Information
Set Membership: |
|

LYRM9
Genomic Map