Assay Details
| Gene Symbol: CDK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714073.3 | XP_002714119.1 | 8 - 9 | 749 | 64 | |
| XM_017344767.1 | XP_017200256.1 | 7 - 8 | 741 | 64 | ||
| XM_017344768.1 | XP_017200257.1 | 6 - 7 | 607 | 64 | ||
More Information
Set Membership: |
|

CDK7
Genomic Map