Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269677.2 | XP_008267899.1 | 3 - 4 | 275 | 65 | |
| XM_008269679.2 | XP_008267901.1 | 3 - 4 | 275 | 65 | ||
| XM_008269680.2 | XP_008267902.1 | 3 - 4 | 276 | 65 | ||
More Information
Set Membership: |
|

GMFB
Genomic Map