Assay Details
| Gene Symbol: SESN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716081.3 | XP_002716127.1 | 4 - 5 | 928 | 55 | |
| XM_017346546.1 | XP_017202035.1 | 3 - 4 | 863 | 55 | ||
More Information
Set Membership: |
|

SESN2
Genomic Map