Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714544.3 | XP_002714590.1 | 6 - 7 | 1490 | 65 | |
| XM_017345248.1 | XP_017200737.1 | 5 - 6 | 1232 | 65 | ||
| XM_017345249.1 | XP_017200738.1 | 5 - 6 | 1232 | 65 | ||
More Information
Set Membership: |
|

LCA5
Genomic Map