Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269938.2 | XP_008268160.1 | 2 - 3 | 580 | 64 | |
| XM_017348696.1 | XP_017204185.1 | 3 - 4 | 711 | 64 | ||
More Information
Set Membership: |
|

AP3M1
Genomic Map