Assay Details
| Gene Symbol: SIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274860.2 | XP_008273082.1 | 1628 | 68 | ||
| XM_017350783.1 | XP_017206272.1 | 1628 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIM2| Gene Symbol: SIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274860.2 | XP_008273082.1 | 1628 | 68 | ||
| XM_017350783.1 | XP_017206272.1 | 1628 | 68 | |||
Set Membership: |
|