Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347343.1 | XP_017202832.1 | 14 - 15 | 4378 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZGRF1| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347343.1 | XP_017202832.1 | 14 - 15 | 4378 | 78 | |
Set Membership: |
|