Assay Details
| Gene Symbol: TMEM165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717081.3 | XP_002717127.1 | 5 - 6 | 1054 | 72 | |
| XM_008267854.2 | XP_008266076.1 | 5 - 6 | 951 | 72 | ||
More Information
Set Membership: |
|

TMEM165
Genomic Map